siRNA Details
| Gene Symbol: 9330198I05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020394.1 | - | 2 | 115 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY721536.1 | - | 2 | 115 | Chr. 5 - 108360187 - 108363382 [+] on Build GRCm38 | |

9330198I05Rik
Genomic Map