siRNA Details
| Gene Symbol: 9330182L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020391.1 | - | 1 | 261 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY721475.1 | - | 1 | 261 | Chr. 12 - 3242281 - 3242906 [+] on Build GRCm38 | |

9330182L19Rik
Genomic Map