siRNA Details
| Gene Symbol: 1700082M22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006980.1 | - | 3 | 1906 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU847445.1 | - | 286 | Chr. - - [-] on Build | ||

1700082M22Rik
Genomic Map