siRNA Details
| Gene Symbol: Jmjd1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133016.1 | - | 2 | 1076 | ||
| AK160952.1 | - | 3 | 1273 | |||
| AK170460.1 | - | 1 | 4504 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jmjd1c| Gene Symbol: Jmjd1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133016.1 | - | 2 | 1076 | ||
| AK160952.1 | - | 3 | 1273 | |||
| AK170460.1 | - | 1 | 4504 | |||