siRNA Details
| Gene Symbol: Ptrf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009381.1 | - | 1 | 1141 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV091925.1 | - | 1 | 268 | Chr. 11 - 100970773 - 100971049 [+] on Build GRCm38 | |

Ptrf
Genomic Map