siRNA Details
| Gene Symbol: Cd47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018679.1 | - | 215 | |||
| AK050387.1 | - | 896 | ||||
| AK164165.1 | - | 2647 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd47| Gene Symbol: Cd47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018679.1 | - | 215 | |||
| AK050387.1 | - | 896 | ||||
| AK164165.1 | - | 2647 | ||||