siRNA Details
| Gene Symbol: Cd47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050387.1 | - | 120 | |||
| AK164165.1 | - | 1871 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CF903449.1 | - | 1 | 557 | Chr. 16 - 49912761 - 49913369 [+] on Build GRCm38 | |

Cd47
Genomic Map