siRNA Details
| Gene Symbol: Gm32511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110517.1 | 5 | 1306 | |||
| GenBank mRNA | AK076745.1 | - | 5 | 1308 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm32511| Gene Symbol: Gm32511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110517.1 | 5 | 1306 | |||
| GenBank mRNA | AK076745.1 | - | 5 | 1308 | ||