siRNA Details
| Gene Symbol: Gm32511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110517.1 | 3, 4 | 1077 | |||
| GenBank mRNA | AK076745.1 | - | 3, 4 | 1079 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm32511| Gene Symbol: Gm32511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110517.1 | 3, 4 | 1077 | |||
| GenBank mRNA | AK076745.1 | - | 3, 4 | 1079 | ||