siRNA Details
| Gene Symbol: Dera | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008033.1 | - | 2 | 261 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY708021.1 | - | 2 | 261 | Chr. 6 - 137754546 - 137760963 [+] on Build GRCm38 | |

Dera
Genomic Map