siRNA Details
| Gene Symbol: 4933425E08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016911.1 | - | 1 | 1000 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB693125.1 | - | 1 | 116 | Chr. 10 - 75835976 - 75836508 [-] on Build GRCm38 | |

4933425E08Rik
Genomic Map