siRNA Details
| Gene Symbol: Gm26994 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK082412.1 | - | 6 | 1091 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm26994| Gene Symbol: Gm26994 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK082412.1 | - | 6 | 1091 | ||