siRNA Details
| Gene Symbol: Gm53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK078566.1 | - | 2 | 351 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB623715.1 | - | 2 | 348 | Chr. 11 - 96251771 - 96262232 [+] on Build GRCm38 | |

Gm53
Genomic Map