siRNA Details
| Gene Symbol: 2010310C07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028457.1 | - | 32 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB612238.1 | - | 1 | 32 | Chr. 6 - 42377956 - 42379658 [+] on Build GRCm38 | |

2010310C07Rik
Genomic Map