siRNA Details
| Gene Symbol: AU015791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139027.1 | - | 1, 2 | 977 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV381755.2 | - | 2, 3 | 190 | Chr. 12 - 105513734 - 105515246 [+] on Build GRCm38 | |

AU015791
Genomic Map