siRNA Details
| Gene Symbol: Khnyn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076856.1 | - | 1 | 1693 | ||
| AK122247.1 | - | 8 | 4209 | |||
| BC034330.1 | - | 1 | 1176 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Khnyn| Gene Symbol: Khnyn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076856.1 | - | 1 | 1693 | ||
| AK122247.1 | - | 8 | 4209 | |||
| BC034330.1 | - | 1 | 1176 | |||