siRNA Details
| Gene Symbol: 1810044D09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038153.1 | 2 | 330 | |||
| GenBank mRNA | AK007770.1 | - | 2 | 332 | ||
| BX529012.1 | - | 2 | 314 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1810044D09Rik| Gene Symbol: 1810044D09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038153.1 | 2 | 330 | |||
| GenBank mRNA | AK007770.1 | - | 2 | 332 | ||
| BX529012.1 | - | 2 | 314 | |||