siRNA Details
Gene Symbol: 1810044D09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038153.1 | 1 | 154 | |||
GenBank mRNA | AK007770.1 | - | 1 | 156 | ||
BX529012.1 | - | 1 | 138 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1810044D09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038153.1 | 1 | 154 | |||
GenBank mRNA | AK007770.1 | - | 1 | 156 | ||
BX529012.1 | - | 1 | 138 |