siRNA Details
| Gene Symbol: 4933424G06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007119.1 | - | 1 | 342 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707198.1 | - | 1 | 339 | Chr. 1 - 36714549 - 36714938 [+] on Build GRCm38 | |

4933424G06Rik
Genomic Map