siRNA Details
Gene Symbol: 4933424G06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040290.1 | 1 | 64 | |||
GenBank mRNA | AK007119.1 | - | 1 | 56 | ||
AK016893.1 | - | 1 | 64 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4933424G06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040290.1 | 1 | 64 | |||
GenBank mRNA | AK007119.1 | - | 1 | 56 | ||
AK016893.1 | - | 1 | 64 |