siRNA Details
| Gene Symbol: 4933424G06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040290.1 | 1 | 22 | |||
| GenBank mRNA | AK007119.1 | - | 1 | 14 | ||
| AK016893.1 | - | 1 | 22 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933424G06Rik| Gene Symbol: 4933424G06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040290.1 | 1 | 22 | |||
| GenBank mRNA | AK007119.1 | - | 1 | 14 | ||
| AK016893.1 | - | 1 | 22 | |||