siRNA Details
| Gene Symbol: 1810059H22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079377.1 | - | 3, 4 | 610 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA198985.1 | - | 2, 3 | 273 | Chr. 5 - 148954445 - 148956601 [-] on Build GRCm38 | |

1810059H22Rik
Genomic Map