siRNA Details
| Gene Symbol: 1810059H22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079377.1 | - | 1 | 246 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB629083.1 | - | 1 | 241 | Chr. 5 - 148954603 - 148960206 [-] on Build GRCm38 | |

1810059H22Rik
Genomic Map