siRNA Details
| Gene Symbol: Baz2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079809.1 | - | 3 | 286 | ||
| AK166079.1 | - | 3 | 274 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Baz2b| Gene Symbol: Baz2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079809.1 | - | 3 | 286 | ||
| AK166079.1 | - | 3 | 274 | |||