siRNA Details
| Gene Symbol: 1700039I01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006642.1 | - | 1 | 460 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706632.1 | - | 1 | 460 | Chr. 1 - 63248021 - 63248499 [-] on Build GRCm38 | |

1700039I01Rik
Genomic Map