siRNA Details
| Gene Symbol: Ggcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030779.1 | - | 2236 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV311526.2 | - | 1 | 951 | Chr. 6 - 72431314 - 72432305 [+] on Build GRCm38 | |

Ggcx
Genomic Map