siRNA Details
| Gene Symbol: Gm15614 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136552.1 | - | 4 | 572 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ054039.1 | - | 4 | 572 | Chr. 5 - 32508099 - 32583386 [-] on Build GRCm38 | |

Gm15614
Genomic Map