siRNA Details
| Gene Symbol: 1700015H07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005995.1 | - | 1 | 314 | ||
| AK076971.1 | - | 1 | 314 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700015H07Rik| Gene Symbol: 1700015H07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005995.1 | - | 1 | 314 | ||
| AK076971.1 | - | 1 | 314 | |||