siRNA Details
| Gene Symbol: 4930487D11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046191.1 | 3 | 313 | |||
| GenBank mRNA | AK019659.1 | - | 3 | 316 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715193.1 | - | 3 | 316 | Chr. 5 - 38344389 - 38349199 [+] on Build GRCm38 | |

4930487D11Rik
Genomic Map