siRNA Details
Gene Symbol: 4930487D11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046191.1 | 1 | 77 | |||
GenBank mRNA | AK019659.1 | - | 1 | 80 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715193.1 | - | 1 | 80 | Chr. 5 - 38344389 - 38349199 [+] on Build GRCm38 |
