siRNA Details
| Gene Symbol: A930019H14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020878.1 | - | 1 | 565 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY742225.1 | - | 1 | 565 | Chr. 10 - 73887453 - 73888115 [+] on Build GRCm38 | |

A930019H14Rik
Genomic Map