siRNA Details
| Gene Symbol: A930019D19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040619.1 | 4 | 523 | |||
| GenBank mRNA | AK020875.1 | - | 4 | 526 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930019D19Rik| Gene Symbol: A930019D19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040619.1 | 4 | 523 | |||
| GenBank mRNA | AK020875.1 | - | 4 | 526 | ||