siRNA Details
| Gene Symbol: 1700025F22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006335.1 | - | 7 | 640 | ||
| BC048529.1 | - | 6 | 567 | 6774736 | ||
| BC061241.1 | - | 7 | 647 | 6743876 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA466362.1 | - | 6 | 570 | Chr. 19 - 11139664 - 11165306 [-] on Build GRCm38 | |

1700025F22Rik
Genomic Map