siRNA Details
| Gene Symbol: 1700025F22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC048529.1 | - | 2 | 139 | 6774736 | |
| BC061241.1 | - | 3 | 219 | 6743876 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700025F22Rik| Gene Symbol: 1700025F22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC048529.1 | - | 2 | 139 | 6774736 | |
| BC061241.1 | - | 3 | 219 | 6743876 | ||