siRNA Details
| Gene Symbol: 4930509E16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045735.1 | 3 | 1060 | |||
| GenBank mRNA | AK015733.1 | - | 3 | 1060 | ||
| BB016321.1 | - | 1 | 115 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930509E16Rik| Gene Symbol: 4930509E16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045735.1 | 3 | 1060 | |||
| GenBank mRNA | AK015733.1 | - | 3 | 1060 | ||
| BB016321.1 | - | 1 | 115 | |||