siRNA Details
| Gene Symbol: 2810433D01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033474.1 | 4 | 567 | |||
| GenBank mRNA | AK019272.1 | - | 4 | 498 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2810433D01Rik| Gene Symbol: 2810433D01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033474.1 | 4 | 567 | |||
| GenBank mRNA | AK019272.1 | - | 4 | 498 | ||