siRNA Details
| Gene Symbol: 2810433D01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033474.1 | 4 | 617 | |||
| GenBank mRNA | AK019272.1 | - | 4 | 548 | ||
| CX565797.1 | - | 1 | 69 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA185245.1 | - | 4 | 530 | Chr. 11 - 102620202 - 102624328 [-] on Build GRCm38 | |

2810433D01Rik
Genomic Map