siRNA Details
Gene Symbol: 4930401O10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045942.1 | 2 | 354 | |||
GenBank mRNA | AK015039.1 | - | 2 | 355 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930401O10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045942.1 | 2 | 354 | |||
GenBank mRNA | AK015039.1 | - | 2 | 355 |