siRNA Details
| Gene Symbol: 1700047G03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC027576.1 | - | 76 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AI019571.1 | - | 1 | 88 | Chr. 15 - 11964337 - 11969524 [+] on Build GRCm38 | |

1700047G03Rik
Genomic Map