siRNA Details
| Gene Symbol: 4930401C12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015031.1 | - | 1 | 219 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU939059.1 | - | 1 | 206 | Chr. 11 - 112156866 - 112157336 [-] on Build GRCm38 | |

4930401C12Rik
Genomic Map