siRNA Details
| Gene Symbol: Gm16006 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042803.1 | - | 2 | 353 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB640009.1 | - | 2 | 353 | Chr. 15 - 57708203 - 57708927 [-] on Build GRCm38 | |

Gm16006
Genomic Map