siRNA Details
| Gene Symbol: Gm4872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073372.1 | 3 | 323 | |||
| GenBank mRNA | AK043311.1 | - | 3 | 324 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB640765.1 | - | 3 | 322 | Chr. 6 - 53221803 - 53226601 [+] on Build GRCm38 | |

Gm4872
Genomic Map