siRNA Details
| Gene Symbol: D430019H16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK052429.1 | - | 2 | 3919 | ||
| AK081558.1 | - | 1 | 993 | |||
| BC058677.1 | - | 1 | 3236 | |||
| BC059046.1 | - | 1 | 215 | |||
| BC078436.1 | - | 1 | 155 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D430019H16Rik| Gene Symbol: D430019H16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK052429.1 | - | 2 | 3919 | ||
| AK081558.1 | - | 1 | 993 | |||
| BC058677.1 | - | 1 | 3236 | |||
| BC059046.1 | - | 1 | 215 | |||
| BC078436.1 | - | 1 | 155 | |||