siRNA Details
| Gene Symbol: D430019H16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK052429.1 | - | 2 | 4567 | ||
| AK081558.1 | - | 1 | 1641 | |||
| BC058677.1 | - | 1 | 3884 | |||
| BC059046.1 | - | 1 | 863 | |||
| BC078436.1 | - | 1 | 803 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D430019H16Rik| Gene Symbol: D430019H16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK052429.1 | - | 2 | 4567 | ||
| AK081558.1 | - | 1 | 1641 | |||
| BC058677.1 | - | 1 | 3884 | |||
| BC059046.1 | - | 1 | 863 | |||
| BC078436.1 | - | 1 | 803 | |||