siRNA Details
| Gene Symbol: Tmem246 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038607.1 | - | 1 | 1026 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI180689.1 | - | 1 | 624 | Chr. 4 - 49596730 - 49597366 [-] on Build GRCm38 | |

Tmem246
Genomic Map