siRNA Details
| Gene Symbol: 6430710M23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018303.1 | - | 1 | 522 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY719191.1 | - | 1 | 522 | Chr. 8 - 25744689 - 25745580 [-] on Build GRCm38 | |

6430710M23Rik
Genomic Map