siRNA Details
| Gene Symbol: 9130403I23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018659.1 | - | 1 | 650 | ||
| AK034835.1 | - | 1 | 677 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9130403I23Rik| Gene Symbol: 9130403I23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018659.1 | - | 1 | 650 | ||
| AK034835.1 | - | 1 | 677 | |||