siRNA Details
| Gene Symbol: 2900054C01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013682.1 | - | 1 | 1229 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY592462.1 | - | 1 | 336 | Chr. X - 160801394 - 160801857 [-] on Build GRCm38 | |

2900054C01Rik
Genomic Map