siRNA Details
| Gene Symbol: 2900054C01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013682.1 | - | 1 | 879 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ261452.1 | - | 1 | 217 | Chr. X - 160801629 - 160802090 [-] on Build GRCm38 | |

2900054C01Rik
Genomic Map