siRNA Details
| Gene Symbol: Palld | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076423.1 | - | 19 | 3483 | ||
| AK077756.1 | - | 1 | 149 | |||
| BC127081.1 | - | 13 | 2617 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Palld| Gene Symbol: Palld | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076423.1 | - | 19 | 3483 | ||
| AK077756.1 | - | 1 | 149 | |||
| BC127081.1 | - | 13 | 2617 | |||